Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113695.1 | NP_001107167.1 | 2 - 3 | 441 | 86 | |
GenBank mRNA | AB215120.1 | - | 2 - 3 | 369 | 86 | |
AF464046.2 | - | 2 - 3 | 362 | 86 | ||
AK348716.1 | - | 2 - 3 | 426 | 86 | ||
AK394000.1 | - | 2 - 3 | 434 | 86 | ||
AK396034.1 | - | 2 - 3 | 433 | 86 | ||
AK396070.1 | - | 2 - 3 | 437 | 86 | ||
AY135576.1 | - | 1 - 2 | 86 | |||
AY135579.1 | - | 2 - 3 | 441 | 86 | ||
DQ883227.1 | - | 2 - 3 | 390 | 86 | ||
EU432072.1 | - | 2 - 3 | 371 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |