Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113704.1 | NP_001107176.1 | 1 - 2 | 112 | 104 | |
XM_013977620.2 | XP_013833074.2 | 170 | 104 | |||
GenBank mRNA | AF464006.1 | - | 1 - 2 | 112 | 104 | |
AF464007.1 | - | 1 - 2 | 112 | 104 | ||
AF464008.1 | - | 1 - 2 | 112 | 104 | ||
AF464019.1 | - | 1 - 2 | 112 | 104 | ||
AF464020.2 | - | 1 - 2 | 74 | 104 | ||
AK236916.1 | - | 1 - 2 | 108 | 104 | ||
AK239694.1 | - | 1 - 2 | 93 | 104 | ||
AK348712.1 | - | 1 - 2 | 97 | 104 | ||
AK348801.1 | - | 1 - 2 | 97 | 104 | ||
AK392987.1 | - | 1 - 2 | 97 | 104 | ||
AK394974.1 | - | 1 - 2 | 97 | 104 | ||
AY247771.1 | - | 1 - 2 | 112 | 104 | ||
AY247772.1 | - | 1 - 2 | 112 | 104 | ||
AY459304.1 | - | 1 - 2 | 112 | 104 | ||
AY463540.1 | - | 1 - 2 | 72 | 104 | ||
DQ883214.1 | - | 1 - 2 | 110 | 104 | ||
DQ883215.1 | - | 1 - 2 | 112 | 104 | ||
DQ883216.1 | - | 1 - 2 | 112 | 104 | ||
DQ976363.1 | - | 1 - 2 | 100 | 104 | ||
DQ992502.1 | - | 1 - 2 | 100 | 104 | ||
DQ992503.1 | - | 1 - 2 | 100 | 104 | ||
DQ992504.1 | - | 1 - 2 | 100 | 104 | ||
DQ992506.1 | - | 1 - 2 | 100 | 104 | ||
DQ992507.1 | - | 1 - 2 | 100 | 104 | ||
DQ992508.1 | - | 1 - 2 | 100 | 104 | ||
DQ992509.1 | - | 1 - 2 | 100 | 104 | ||
DQ992510.1 | - | 1 - 2 | 100 | 104 | ||
EU432054.1 | - | 1 - 2 | 93 | 104 | ||
EU432055.1 | - | 1 - 2 | 93 | 104 | ||
EU432056.1 | - | 1 - 2 | 93 | 104 | ||
EU432057.1 | - | 1 - 2 | 93 | 104 | ||
GU322911.1 | - | 1 - 2 | 78 | 104 | ||
GU322912.1 | - | 1 - 2 | 78 | 104 | ||
GU322913.1 | - | 1 - 2 | 78 | 104 | ||
GU322914.1 | - | 1 - 2 | 78 | 104 | ||
GU322915.1 | - | 1 - 2 | 78 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |