Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114284.2 | NP_001107756.1 | 4 - 5 | 370 | 91 | |
GenBank mRNA | AK237194.1 | - | 736 | 91 | ||
AK350182.1 | - | 4 - 5 | 428 | 91 | ||
AK391704.1 | - | 4 - 5 | 403 | 91 | ||
AK393844.1 | - | 4 - 5 | 402 | 91 | ||
AK394513.1 | - | 4 - 5 | 408 | 91 | ||
AK398700.1 | - | 4 - 5 | 404 | 91 | ||
EU352798.1 | - | 4 - 5 | 371 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |