Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122983.1 | NP_001116455.1 | 1 - 2 | 110 | 70 | |
XM_021085889.1 | XP_020941548.1 | 552 | 70 | |||
GenBank mRNA | AB529865.1 | - | 1 - 2 | 98 | 70 | |
AK235636.1 | - | 2 - 3 | 156 | 70 | ||
EU534186.1 | - | 1 - 2 | 110 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |