Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122983.1 | NP_001116455.1 | 4 - 5 | 592 | 87 | |
XM_021085889.1 | XP_020941548.1 | 1034 | 87 | |||
GenBank mRNA | AB529865.1 | - | 4 - 5 | 580 | 87 | |
AK235636.1 | - | 5 - 6 | 638 | 87 | ||
EU534186.1 | - | 4 - 5 | 592 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |