Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123043.1 | NP_001116515.1 | 1 - 2 | 158 | 86 | |
XM_005665245.3 | XP_005665302.1 | 347 | 86 | |||
GenBank mRNA | AK390132.1 | - | 2 - 3 | 294 | 86 | |
AK400244.1 | - | 2 - 3 | 326 | 86 | ||
DQ917651.1 | - | 1 - 2 | 158 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |