Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123111.1 | NP_001116583.1 | 2 - 3 | 178 | 109 | |
XM_013977649.2 | XP_013833103.1 | 709 | 109 | |||
XM_013977651.2 | XP_013833105.1 | 711 | 109 | |||
XM_021097962.1 | XP_020953621.1 | 517 | 109 | |||
XM_021097963.1 | XP_020953622.1 | 707 | 109 | |||
GenBank mRNA | EU282362.1 | - | 2 - 3 | 190 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |