Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123143.1 | NP_001116615.1 | 6 - 7 | 754 | 78 | |
XM_005664831.3 | XP_005664888.1 | 1437 | 78 | |||
GenBank mRNA | AB516285.1 | - | 6 - 7 | 784 | 78 | |
AK396179.1 | - | 6 - 7 | 783 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |