Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123157.1 | NP_001116629.1 | 2 - 3 | 568 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |