Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss03390704_m1
See other RHOF GE Assays ›
Gene Symbol:
RHOF
Gene Name:
ras homolog family member F, filopodia associated
Gene Aliases:
-
Chromosome Location:
Chr.14: 30886053 - 30908048 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss03390704_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: RHOF
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001123190.1
NP_001116662.1
4 - 5
472
65
XM_013982810.2
XP_013838264.1
5094
65
XM_021071816.1
XP_020927475.1
5182
65
XM_021071817.1
XP_020927476.1
3596
65
XM_021071818.1
XP_020927477.1
3707
65
XM_021071819.1
XP_020927478.1
3500
65
XM_021071820.1
XP_020927479.1
3924
65
XM_021071821.1
XP_020927480.1
4272
65
XM_021071822.1
XP_020927481.1
3634
65
XM_021071823.1
XP_020927482.1
3347
65
XM_021071825.1
XP_020927484.1
3458
65
XM_021071826.1
XP_020927485.1
3772
65
XM_021071827.1
XP_020927486.1
3383
65
XM_021071828.1
XP_020927487.1
3773
65
XM_021071829.1
XP_020927488.1
3611
65
XM_021071830.1
XP_020927489.1
3470
65
XM_021071831.1
XP_020927490.1
3750
65
XM_021071832.1
XP_020927491.1
3340
65
XM_021071833.1
XP_020927492.1
4173
65
XM_021071834.1
XP_020927493.1
3944
65
XM_021071835.1
XP_020927494.1
3703
65
XM_021071836.1
XP_020927495.1
3458
65
XM_021071837.1
XP_020927496.1
3362
65
XM_021071838.1
XP_020927497.1
3871
65
XM_021071839.1
XP_020927498.1
3786
65
XM_021071840.1
XP_020927499.1
4317
65
XM_021071841.1
XP_020927500.1
3496
65
XM_021071842.1
XP_020927501.1
3209
65
XM_021071843.1
XP_020927502.1
4170
65
XM_021071844.1
XP_020927503.1
3860
65
XM_021071845.1
XP_020927504.1
3118
65
XM_021071846.1
XP_020927505.1
3356
65
XM_021071847.1
XP_020927506.1
3108
65
XM_021071848.1
XP_020927507.1
3063
65
XM_021071849.1
XP_020927508.1
3210
65
XM_021071850.1
XP_020927509.1
4179
65
XM_021071851.1
XP_020927510.1
3756
65
XM_021071852.1
XP_020927511.1
3365
65
XM_021071853.1
XP_020927512.1
3467
65
XM_021071854.1
XP_020927513.1
3259
65
XM_021071856.1
XP_020927515.1
3107
65
XM_021071857.1
XP_020927516.1
6409
65
XM_021071858.1
XP_020927517.1
2884
65
XM_021071859.1
XP_020927518.1
3071
65
XM_021071860.1
XP_020927519.1
3370
65
XM_021071861.1
XP_020927520.1
3443
65
XM_021071862.1
XP_020927521.1
688
65
XM_021071863.1
XP_020927522.1
5173
65
XM_021071864.1
XP_020927523.1
3386
65
GenBank mRNA
DQ917638.1
-
4 - 5
472
65
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
small GTPase
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0