Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123201.1 | NP_001116673.1 | 4 - 5 | 1049 | 75 | |
GenBank mRNA | AK349972.1 | - | 4 - 5 | 1037 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |