Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNXB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123204.1 | NP_001116676.1 | 2928 | 68 | ||
XM_021098334.1 | XP_020953993.1 | 2904 | 68 | |||
XM_021098340.1 | XP_020953999.1 | 2904 | 68 |