Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123213.1 | NP_001116685.1 | 2 - 3 | 550 | 134 | |
XM_021077209.1 | XP_020932868.1 | 7722 | 134 | |||
GenBank mRNA | AK396657.1 | - | 2 - 3 | 467 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |