Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123216.1 | NP_001116688.1 | 2 - 3 | 388 | 69 | |
GenBank mRNA | AK239309.1 | - | 2 - 3 | 69 | ||
AK350043.1 | - | 2 - 3 | 69 | |||
FJ455510.1 | - | 2 - 3 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |