Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123222.1 | NP_001116694.1 | 8 - 9 | 1096 | 100 | |
GenBank mRNA | EU442573.1 | - | 8 - 9 | 1096 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |