Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213922.1 | NP_999087.1 | 7 - 8 | 812 | 110 | |
XM_021072076.1 | XP_020927735.1 | 933 | 110 | |||
GenBank mRNA | AY248702.1 | - | 7 - 8 | 812 | 110 | |
AY609421.1 | - | 7 - 8 | 805 | 110 | ||
FJ475066.1 | - | 7 - 8 | 812 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |