Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP27B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213995.1 | NP_999160.1 | 5 - 6 | 988 | 85 | |
XM_021091124.1 | XP_020946783.1 | 1383 | 85 | |||
GenBank mRNA | AB018743.1 | - | 5 - 6 | 988 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |