Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP91-PHOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214043.2 | NP_999208.1 | 5 - 6 | 641 | 78 | |
GenBank mRNA | AK230821.1 | - | 8 - 9 | 972 | 78 | |
AK231643.1 | - | 8 - 9 | 973 | 78 | ||
AK232201.1 | - | 8 - 9 | 977 | 78 | ||
AK345180.1 | - | 8 - 9 | 973 | 78 | ||
AK398894.1 | - | 8 - 9 | 952 | 78 | ||
AK398895.1 | - | 8 - 9 | 952 | 78 | ||
U02476.1 | - | 5 - 6 | 641 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |