Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214049.1 | NP_999214.1 | 6 - 7 | 1058 | 101 | |
GenBank mRNA | AF095744.1 | - | 6 - 7 | 1058 | 101 | |
AF128837.1 | - | 5 - 6 | 868 | 101 | ||
DQ530368.1 | - | 5 - 6 | 938 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |