Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214083.2 | NP_999248.2 | 3 - 4 | 461 | 69 | |
XM_021079496.1 | XP_020935155.1 | 923 | 69 | |||
XM_021079497.1 | XP_020935156.1 | 733 | 69 | |||
GenBank mRNA | AB040442.1 | - | 3 - 4 | 461 | 69 | |
AK239062.1 | - | 3 - 4 | 494 | 69 | ||
AK390911.1 | - | 3 - 4 | 498 | 69 | ||
EU026383.1 | - | 3 - 4 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |