Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214374.2 | NP_999539.2 | 6 - 7 | 552 | 130 | |
GenBank mRNA | AK347117.1 | - | 6 - 7 | 712 | 130 | |
AK347215.1 | - | 6 - 7 | 878 | 130 | ||
AK347231.1 | - | 6 - 7 | 562 | 130 | ||
AK394636.1 | - | 6 - 7 | 508 | 130 | ||
AK394638.1 | - | 7 - 8 | 536 | 130 | ||
AK394642.1 | - | 6 - 7 | 721 | 130 | ||
AK394655.1 | - | 6 - 7 | 539 | 130 | ||
AK394666.1 | - | 6 - 7 | 539 | 130 | ||
AK394667.1 | - | 6 - 7 | 542 | 130 | ||
AK394722.1 | - | 6 - 7 | 543 | 130 | ||
AK399622.1 | - | 6 - 7 | 541 | 130 | ||
AK399655.1 | - | 6 - 7 | 539 | 130 | ||
AY680668.1 | - | 6 - 7 | 560 | 130 | ||
AY680669.1 | - | 6 - 7 | 560 | 130 | ||
AY864611.1 | - | 6 - 7 | 552 | 130 | ||
AY864615.1 | - | 7 - 8 | 582 | 130 | ||
AY864616.1 | - | 6 - 7 | 521 | 130 | ||
AY894693.1 | - | 6 - 7 | 552 | 130 | ||
DQ629158.1 | - | 6 - 7 | 719 | 130 | ||
DQ629159.1 | - | 6 - 7 | 541 | 130 | ||
DQ629160.1 | - | 7 - 8 | 568 | 130 | ||
GU195782.1 | - | 7 - 8 | 569 | 130 | ||
GU195783.1 | - | 6 - 7 | 510 | 130 | ||
GU195784.1 | - | 6 - 7 | 505 | 130 | ||
Y18404.1 | - | 116 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |