Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001625.2 | NP_001001625.2 | 780 | 87 | ||
GenBank mRNA | AY533208.1 | - | 552 | 87 | ||
HQ316945.1 | - | 552 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa |