Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001630.1 | NP_001001630.1 | 4 - 5 | 317 | 60 | |
XM_013990916.2 | XP_013846370.1 | 287 | 60 | |||
XM_013990917.2 | XP_013846371.1 | 278 | 60 | |||
XM_021079531.1 | XP_020935190.1 | 220 | 60 | |||
GenBank mRNA | AK231668.1 | - | 4 - 5 | 320 | 60 | |
AK345447.1 | - | 4 - 5 | 298 | 60 | ||
AK346609.1 | - | 4 - 5 | 303 | 60 | ||
AY550924.1 | - | 4 - 5 | 317 | 60 | ||
AY609748.1 | - | 4 - 5 | 296 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |