Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001634.1 | NP_001001634.1 | 2 - 3 | 266 | 94 | |
GenBank mRNA | AK239565.1 | - | 1 | 290 | 94 | |
AK352159.1 | - | 1265 | 94 | |||
AY550073.1 | - | 2 - 3 | 266 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |