Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFN-DELTA-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002832.1 | NP_001002832.1 | 1 | 548 | 107 | |
GenBank mRNA | Z22706.1 | - | 548 | 107 |
Gene Symbol: IFN-DELTA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166321.2 | NP_001159793.2 | 1 | 548 | 107 | |
XM_021063028.1 | XP_020918687.1 | 785 | 107 | |||
GenBank mRNA | GQ415074.1 | - | 1 | 341 | 107 | |
GQ415075.1 | - | 1 | 341 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |