Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003926.2 | NP_001003926.1 | 5 - 6 | 944 | 107 | |
GenBank mRNA | AY523565.2 | - | 5 - 6 | 946 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |