Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005150.1 | NP_001005150.1 | 2 - 3 | 340 | 90 | |
GenBank mRNA | AK233732.1 | - | 1 - 2 | 104 | 90 | |
AK239662.1 | - | 3 - 4 | 600 | 90 | ||
AK389460.1 | - | 2 - 3 | 448 | 90 | ||
AK395569.1 | - | 2 - 3 | 716 | 90 | ||
AK397107.1 | - | 2 - 3 | 486 | 90 | ||
AK398938.1 | - | 2 - 3 | 451 | 90 | ||
AK399004.1 | - | 2 - 3 | 465 | 90 | ||
AK399188.1 | - | 2 - 3 | 451 | 90 | ||
AY609599.1 | - | 2 - 3 | 437 | 90 | ||
FN421467.1 | - | 2 - 3 | 277 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |