Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005150.1 | NP_001005150.1 | 3 - 4 | 552 | 61 | |
GenBank mRNA | AK233732.1 | - | 2 - 3 | 316 | 61 | |
AK239662.1 | - | 4 - 5 | 812 | 61 | ||
AK395569.1 | - | 3 - 4 | 928 | 61 | ||
AK395997.1 | - | 2 - 3 | 1014 | 61 | ||
AK397107.1 | - | 3 - 4 | 698 | 61 | ||
AK398938.1 | - | 3 - 4 | 663 | 61 | ||
AK399004.1 | - | 3 - 4 | 677 | 61 | ||
AK399095.1 | - | 2 - 3 | 557 | 61 | ||
AK399188.1 | - | 3 - 4 | 663 | 61 | ||
AY609599.1 | - | 3 - 4 | 649 | 61 | ||
FN421467.1 | - | 3 - 4 | 489 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |