Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005157.1 | NP_001005157.1 | 2 - 3 | 339 | 73 | |
XM_005665792.3 | XP_005665849.1 | 575 | 73 | |||
XM_005665793.3 | XP_005665850.1 | 562 | 73 | |||
XM_005665794.3 | XP_005665851.1 | 626 | 73 | |||
XM_021098341.1 | XP_020954000.1 | 1006 | 73 | |||
GenBank mRNA | AK236289.1 | - | 3 - 4 | 513 | 73 | |
AK238677.1 | - | 3 - 4 | 528 | 73 | ||
AK389641.1 | - | 3 - 4 | 623 | 73 | ||
AK389706.1 | - | 3 - 4 | 530 | 73 | ||
AK399598.1 | - | 3 - 4 | 530 | 73 | ||
EU282340.1 | - | 2 - 3 | 990 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |