Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005727.1 | NP_001005727.1 | 5 - 6 | 644 | 82 | |
XM_005672834.1 | XP_005672891.1 | 823 | 82 | |||
XM_005672835.2 | XP_005672892.1 | 719 | 82 | |||
GenBank mRNA | AK238135.1 | - | 6 - 7 | 701 | 82 | |
AK238161.1 | - | 6 - 7 | 722 | 82 | ||
AK238256.1 | - | 6 - 7 | 701 | 82 | ||
AK238296.1 | - | 5 - 6 | 1177 | 82 | ||
AK350801.1 | - | 5 - 6 | 688 | 82 | ||
AK350937.1 | - | 6 - 7 | 702 | 82 | ||
AK397439.1 | - | 6 - 7 | 701 | 82 | ||
AK397446.1 | - | 6 - 7 | 701 | 82 | ||
AK397447.1 | - | 6 - 7 | 702 | 82 | ||
AK397448.1 | - | 6 - 7 | 701 | 82 | ||
AK397451.1 | - | 6 - 7 | 701 | 82 | ||
AK397460.1 | - | 6 - 7 | 701 | 82 | ||
AK397478.1 | - | 6 - 7 | 704 | 82 | ||
AK397484.1 | - | 6 - 7 | 701 | 82 | ||
AK397490.1 | - | 6 - 7 | 823 | 82 | ||
AK397496.1 | - | 6 - 7 | 727 | 82 | ||
AK397506.1 | - | 6 - 7 | 701 | 82 | ||
AK397531.1 | - | 5 - 6 | 1202 | 82 | ||
AK397535.1 | - | 6 - 7 | 701 | 82 | ||
AK397538.1 | - | 6 - 7 | 701 | 82 | ||
AK397555.1 | - | 6 - 7 | 707 | 82 | ||
AK397556.1 | - | 6 - 7 | 701 | 82 | ||
AK397606.1 | - | 6 - 7 | 701 | 82 | ||
AK397619.1 | - | 6 - 7 | 705 | 82 | ||
AK397620.1 | - | 6 - 7 | 706 | 82 | ||
AK397956.1 | - | 6 - 7 | 707 | 82 | ||
AK397957.1 | - | 6 - 7 | 703 | 82 | ||
AK397999.1 | - | 6 - 7 | 707 | 82 | ||
AK398049.1 | - | 6 - 7 | 708 | 82 | ||
AK398104.1 | - | 6 - 7 | 703 | 82 | ||
AK399196.1 | - | 6 - 7 | 701 | 82 | ||
AK399197.1 | - | 6 - 7 | 732 | 82 | ||
AK399199.1 | - | 6 - 7 | 704 | 82 | ||
AK399201.1 | - | 6 - 7 | 701 | 82 | ||
AK399203.1 | - | 6 - 7 | 700 | 82 | ||
AK399205.1 | - | 6 - 7 | 701 | 82 | ||
AK399208.1 | - | 6 - 7 | 705 | 82 | ||
AK399209.1 | - | 6 - 7 | 706 | 82 | ||
AK399210.1 | - | 6 - 7 | 705 | 82 | ||
AK399218.1 | - | 6 - 7 | 705 | 82 | ||
AK399221.1 | - | 6 - 7 | 701 | 82 | ||
AK399225.1 | - | 6 - 7 | 711 | 82 | ||
AK399227.1 | - | 6 - 7 | 717 | 82 | ||
AK399228.1 | - | 6 - 7 | 705 | 82 | ||
AK399233.1 | - | 6 - 7 | 701 | 82 | ||
AK399234.1 | - | 6 - 7 | 706 | 82 | ||
AK399236.1 | - | 6 - 7 | 701 | 82 | ||
AK399238.1 | - | 6 - 7 | 701 | 82 | ||
AK399241.1 | - | 6 - 7 | 701 | 82 | ||
AK399242.1 | - | 6 - 7 | 701 | 82 | ||
AK399245.1 | - | 6 - 7 | 701 | 82 | ||
AK399248.1 | - | 6 - 7 | 701 | 82 | ||
AK399252.1 | - | 6 - 7 | 707 | 82 | ||
AK399256.1 | - | 6 - 7 | 703 | 82 | ||
AK399257.1 | - | 6 - 7 | 705 | 82 | ||
AK399259.1 | - | 6 - 7 | 701 | 82 | ||
AK399260.1 | - | 6 - 7 | 709 | 82 | ||
AK399261.1 | - | 6 - 7 | 701 | 82 | ||
AK399334.1 | - | 6 - 7 | 703 | 82 | ||
AK399340.1 | - | 6 - 7 | 704 | 82 | ||
AK399348.1 | - | 6 - 7 | 705 | 82 | ||
AK399350.1 | - | 6 - 7 | 703 | 82 | ||
AK399351.1 | - | 6 - 7 | 711 | 82 | ||
AY733063.1 | - | 5 - 6 | 644 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |