Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025215.1 | NP_001020386.1 | 3 - 4 | 558 | 125 | |
XM_005656299.3 | XP_005656356.1 | 552 | 125 | |||
GenBank mRNA | AK240074.1 | - | 3 - 4 | 481 | 125 | |
AK240576.1 | - | 4 - 5 | 226 | 125 | ||
AK391778.1 | - | 3 - 4 | 469 | 125 | ||
AK396926.1 | - | 3 - 4 | 227 | 125 | ||
AK400614.1 | - | 3 - 4 | 499 | 125 | ||
DQ058293.1 | - | 3 - 4 | 558 | 125 | ||
EU561660.1 | - | 3 - 4 | 449 | 125 | ||
EU561661.1 | - | 3 - 4 | 449 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |