Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025225.1 | NP_001020396.1 | 1 - 2 | 45 | 76 | |
XM_005673034.3 | XP_005673091.1 | 448 | 76 | |||
XM_005673035.3 | XP_005673092.1 | 298 | 76 | |||
XM_005673036.3 | XP_005673093.1 | 298 | 76 | |||
GenBank mRNA | AB196773.1 | - | 1 - 2 | 45 | 76 | |
EF486358.1 | - | 1 - 2 | 60 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |