Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033011.1 | NP_001028183.1 | 3 - 4 | 301 | 65 | |
GenBank mRNA | AK345322.1 | - | 3 - 4 | 352 | 65 | |
AK345770.1 | - | 3 - 4 | 330 | 65 | ||
DQ026063.1 | - | 3 - 4 | 301 | 65 | ||
KF595282.1 | - | 3 - 4 | 300 | 65 | ||
KF595283.1 | - | 3 - 4 | 300 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |