Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033011.1 | NP_001028183.1 | 4 - 5 | 562 | 83 | |
GenBank mRNA | AK345770.1 | - | 4 - 5 | 591 | 83 | |
DQ026063.1 | - | 4 - 5 | 562 | 83 | ||
KF595281.1 | - | 4 - 5 | 561 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |