Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037276.1 | NP_001032353.1 | 1 - 2 | 125 | 72 | |
XM_005672935.3 | XP_005672992.1 | 203 | 72 | |||
XM_013985367.2 | XP_013840821.1 | 211 | 72 | |||
XM_013985368.2 | XP_013840822.1 | 211 | 72 | |||
XM_013985370.2 | XP_013840824.1 | 204 | 72 | |||
GenBank mRNA | DQ191892.1 | - | 1 - 2 | 125 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |