Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037961.1 | NP_001033050.1 | 3 - 4 | 447 | 71 | |
GenBank mRNA | AK231242.1 | - | 3 - 4 | 293 | 71 | |
DQ295031.1 | - | 3 - 4 | 447 | 71 | ||
DQ333333.1 | - | 3 - 4 | 305 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |