Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038638.1 | NP_001033727.1 | 1 - 2 | 67 | 77 | |
GenBank mRNA | AY973170.1 | - | 1 - 2 | 67 | 77 | |
EU416277.1 | - | 1 - 2 | 77 | |||
JQ044413.1 | - | 1 - 2 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |