Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038638.1 | NP_001033727.1 | 2 - 3 | 268 | 71 | |
GenBank mRNA | AY973170.1 | - | 2 - 3 | 268 | 71 | |
EU416277.1 | - | 2 - 3 | 251 | 71 | ||
JQ044413.1 | - | 2 - 3 | 248 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Inventoried Probe spans exons |