Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039746.2 | NP_001034835.1 | 2 - 3 | 326 | 99 | |
GenBank mRNA | AB529462.1 | - | 2 - 3 | 244 | 99 | |
AK231381.1 | - | 2 - 3 | 327 | 99 | ||
AK346851.1 | - | 484 | 99 | |||
AY841270.1 | - | 2 - 3 | 329 | 99 | ||
DQ372083.1 | - | 2 - 3 | 327 | 99 | ||
DQ523617.1 | - | 2 - 3 | 211 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |