Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213760.2 | NP_998925.2 | 2 | 1380 | 75 | |
GenBank mRNA | AB085746.1 | - | 2 | 1377 | 75 | |
AB085936.1 | - | 1 | 1253 | 75 | ||
KF019634.1 | - | 1 | 1253 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |