Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213805.1 | NP_998970.1 | 5 - 6 | 729 | 78 | |
GenBank mRNA | AB018668.1 | - | 5 - 6 | 729 | 78 | |
AK230974.1 | - | 5 - 6 | 799 | 78 | ||
AK343993.1 | - | 5 - 6 | 806 | 78 | ||
EU122440.1 | - | 5 - 6 | 685 | 78 | ||
KJ786421.1 | - | 5 - 6 | 725 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |