Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF313487.1 | - | 3 - 4 | 463 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |