Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213909.1 | NP_999074.1 | 2 | 250 | 98 | |
XM_021102200.1 | XP_020957859.1 | 412 | 98 | |||
XM_021102201.1 | XP_020957860.1 | 552 | 98 | |||
XM_021102202.1 | XP_020957861.1 | 512 | 98 | |||
GenBank mRNA | AB529528.1 | - | 1 | 133 | 98 | |
AK230941.1 | - | 2 | 262 | 98 | ||
AK390323.1 | - | 3 | 384 | 98 | ||
Z29636.1 | - | 250 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |