Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARGC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213963.2 | NP_999128.2 | 12 | 2290 | 96 | |
XM_021100438.1 | XP_020956097.1 | 3062 | 96 | |||
XM_021100439.1 | XP_020956098.1 | 2787 | 96 | |||
XM_021100440.1 | XP_020956099.1 | 5078 | 96 | |||
XM_021100441.1 | XP_020956100.1 | 3183 | 96 | |||
XM_021100442.1 | XP_020956101.1 | 5553 | 96 | |||
XM_021100444.1 | XP_020956103.1 | 4549 | 96 | |||
GenBank mRNA | AB106108.1 | - | 12 | 2290 | 96 | |
AH013165.2 | - | 12 | 2348 | 96 | ||
AK235681.1 | - | 12 | 2427 | 96 | ||
AK393283.1 | - | 13 | 2479 | 96 | ||
AK393759.1 | - | 12 | 2378 | 96 | ||
AK398499.1 | - | 5 | 775 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Inventoried Amplicon spans exons and probe does not span exons |