Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213981.1 | NP_999146.1 | 9 | 848 | 69 | |
GenBank mRNA | AK231202.1 | - | 8 | 831 | 69 | |
AK231584.1 | - | 9 | 883 | 69 | ||
AK344919.1 | - | 8 | 828 | 69 | ||
AK391165.1 | - | 9 | 1092 | 69 | ||
AK392971.1 | - | 9 | 860 | 69 | ||
AK393040.1 | - | 8 | 863 | 69 | ||
AK398839.1 | - | 8 | 836 | 69 | ||
X79303.1 | - | 848 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |