Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214070.1 | NP_999235.1 | 10 - 11 | 1166 | 76 | |
GenBank mRNA | U43490.1 | - | 10 - 11 | 1166 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |