Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214110.1 | NP_999275.1 | 3 - 4 | 364 | 67 | |
XM_005671145.3 | XP_005671202.1 | 582 | 67 | |||
XM_013983376.2 | XP_013838830.1 | 411 | 67 | |||
GenBank mRNA | AF132496.2 | - | 3 - 4 | 364 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |