Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214133.1 | NP_999298.1 | 1 - 2 | 233 | 89 | |
GenBank mRNA | AF350911.2 | - | 1 - 2 | 233 | 89 | |
AK235021.1 | - | 1 - 2 | 232 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |