Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214147.2 | NP_999312.2 | 1 - 2 | 244 | 80 | |
GenBank mRNA | AK231283.1 | - | 1 - 2 | 244 | 80 | |
AK344401.1 | - | 1 - 2 | 233 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |