Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214186.1 | NP_999351.1 | 1 - 2 | 393 | 147 | |
GenBank mRNA | AY081951.1 | - | 1 - 2 | 393 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |